Assay Details
Target Gene Details
Entrez Gene ID: | 212377 |
Gene Name: | MMS22-like, DNA repair protein |
Gene Aliases: |
F730047E07Rik, Gm134 |
Location: |
Chr.4:24496444-24602950 on Build GRCm38 |
Assay Gene Location: | Within Exon 22 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Mms22l | NM_199467.2 | 22 | 3870 | NP_955761.2 |
| AK089531.1 | 14 | 2399 | ||
| AK144799.1 | 21 | 3670 | ||
| AY512923.1 | 17 | 3007 | AAR87794.1 | |
| BC025516.1 | 5 | 778 | AAH25516.1 | |
| BC051674.1 | 13 | 2317 | AAH51674.1 | |
| BC137910.1 | 17 | 2780 | ||
| BC145028.1 | 17 | 2780 |

Genomic Map