Assay Details
Target Gene Details
Entrez Gene ID: | 212377 |
Gene Name: | MMS22-like, DNA repair protein |
Gene Aliases: |
F730047E07Rik, Gm134 |
Location: |
Chr.4:24496444-24602950 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 2 - Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Mms22l | NM_199467.2 | NP_955761.2 | ||
| AK144799.1 | ||||
| AK164069.1 | BAE37613.1 | |||
| AK166072.1 | ||||
| CK639437.1 |
More Information
Set Membership: |
|

Genomic Map