Assay Details
Target Gene Details
Entrez Gene ID: | 212377 |
Gene Name: | MMS22-like, DNA repair protein |
Gene Aliases: |
F730047E07Rik, Gm134 |
Location: |
Chr.4:24496444-24602950 on Build GRCm38 |
Assay Gene Location: | Within Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Mms22l | NM_199467.2 | 9 | 1508 | NP_955761.2 |
| AK089531.1 | 2 | 248 | ||
| AK144799.1 | 9 | 1427 | ||
| AK166072.1 | 9 | 1519 | ||
| AY512923.1 | 4 | 642 | AAR87794.1 | |
| BC137910.1 | 4 | 418 | ||
| BC145028.1 | 4 | 418 |

Genomic Map