Assay Details
Target Gene Details
Entrez Gene ID: | 319247 |
Gene Name: | RIKEN cDNA 9530080O11 gene |
Gene Aliases: |
- |
Location: |
Chr.4:95959711-95967655 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| 9530080O11Rik | NR_045776.1 | 1 | 113 | |
| AK035647.1 | 1 | 113 | BAC29137.1 | |
| AK079280.1 | 1 | 113 | BAC37595.1 |
Target Gene Details
Entrez Gene ID: | 77963 |
Gene Name: | hook homolog 1 (Drosophila) |
Gene Aliases: |
A930033L17Rik, azh |
Location: |
Chr.4:95967187-96024277 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 1 - Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Hook1 | NM_030014.2 | NP_084290.1 | ||
| AF487912.1 | AAM74055.1 | |||
| AK020924.1 | BAB32257.1 | |||
| AK028714.1 | ||||
| AK038941.1 | BAC30175.1 | |||
| AK043870.1 | BAC31686.1 | |||
| AK049897.1 | BAC33977.1 | |||
| BC061688.1 | AAH61688.1 |
More Information
Set Membership: |
|

Genomic Map