Assay Details
Target Gene Details
Entrez Gene ID: | 54366 |
Gene Name: | catenin (cadherin associated protein), alpha-like 1 |
Gene Aliases: |
ACRP, AI616177, AW545119, C86009, Catnal1 |
Location: |
Chr.4:56810935-56865211 on Build GRCm38 |
Assay Gene Location: | Within Exon 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ctnnal1 | NM_018761.3 | 10 | 1492 | NP_061231.3 |
| AF006071.1 | 10 | 1479 | AAC26012.1 | |
| AK034393.1 | 11 | 1492 | ||
| AK038548.1 | 7 | 1429 | ||
| AK082755.1 | 10 | 1436 | ||
| BC050070.1 | 10 | 1469 | AAH50070.1 |

Genomic Map