Assay Details
Target Gene Details
Entrez Gene ID: | 50798 |
Gene Name: | glucosamine (UDP-N-acetyl)-2-epimerase/N-acetylmannosamine kinase |
Gene Aliases: |
2310066H07Rik, DMRV, GLCNE, IBM2, NM, Uae1 |
Location: |
Chr.4:44034075-44084177 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 4 - Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Gne | NM_001190414.1 | NP_001177343.1 | ||
| NM_015828.3 | NP_056643.3 | |||
| AJ132236.1 | CAB36908.1 | |||
| AK136573.1 | BAE23055.1 | |||
| AK170707.1 | BAE41968.1 | |||
| AK197576.1 | ||||
| AK202795.1 | ||||
| AK219474.1 | ||||
| AM697710.1 | ||||
| BC015277.1 | AAH15277.1 | |||
| BC051254.1 | AAH51254.1 |
More Information
Set Membership: |
|

Genomic Map