Assay Details
Target Gene Details
Entrez Gene ID: | 50798 |
Gene Name: | glucosamine (UDP-N-acetyl)-2-epimerase/N-acetylmannosamine kinase |
Gene Aliases: |
2310066H07Rik, DMRV, GLCNE, IBM2, NM, Uae1 |
Location: |
Chr.4:44034075-44084177 on Build GRCm38 |
Assay Gene Location: | Within Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Gne | NM_001190414.1 | 5 | 1200 | NP_001177343.1 |
| NM_015828.3 | 5 | 1018 | NP_056643.3 | |
| AJ132236.1 | 4 | 846 | CAB36908.1 | |
| AK010070.1 | 2 | 166 | ||
| AK136573.1 | 5 | 1019 | BAE23055.1 | |
| AK170707.1 | 5 | 1201 | BAE41968.1 | |
| AM697710.1 | 5 | 939 | ||
| BC015277.1 | 5 | 1006 | AAH15277.1 | |
| BC051254.1 | 5 | 1006 | AAH51254.1 |
More Information
Set Membership: |
|

Genomic Map