Assay Details
Target Gene Details
Entrez Gene ID: | 30952 |
Gene Name: | cyclic nucleotide gated channel beta 3 |
Gene Aliases: |
CCNC2, CNG6, Cngbeta2 |
Location: |
Chr.4:19280850-19510623 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 6 - Intron 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Cngb3 | NM_013927.2 | NP_038955.1 | ||
| AJ243572.1 | CAB71152.1 |
More Information
Set Membership: |
|

Genomic Map