Assay Details
Target Gene Details
Entrez Gene ID: | 320790 |
Gene Name: | chromodomain helicase DNA binding protein 7 |
Gene Aliases: |
A730019I05Rik, Cycn, Cyn, Dz, Edy, Flo, GENA 47, GENA 60, Gena 52, Lda, Mt, Obt, Todo, WBE1, Whi, metis |
Location: |
Chr.4:8690406-8868449 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 19 - Intron 19 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Chd7 | NM_001277149.1 | NP_001264078.1 |

Genomic Map