Assay Details
Target Gene Details
Entrez Gene ID: | 76299 |
Gene Name: | endoplasmic reticulum protein 44 |
Gene Aliases: |
1110001E24Rik, AI849526, AL033348, Txndc4 |
Location: |
Chr.4:48193331-48279601 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 11 - Exon 11 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Erp44 | NM_029572.2 | NP_083848.1 | ||
| AK003217.1 | BAB22648.1 | |||
| AK141377.1 | BAE24664.1 | |||
| AK151497.1 | BAE30449.1 | |||
| AK160113.1 | BAE35637.1 | |||
| AK190947.1 | ||||
| AK206483.1 | ||||
| AK214712.1 | ||||
| BC019558.1 | AAH19558.1 |

Genomic Map