Assay Details
Target Gene Details
Entrez Gene ID: | 654318 |
Gene Name: | RIKEN cDNA C530005A16 gene |
Gene Aliases: |
D930044c15rik |
Location: |
Chr.4:116589733-116597630 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| C530005A16Rik | NR_029450.1 | 3 | 1981 | |
| AK086654.1 | 3 | 1983 | BAC39711.1 |
Target Gene Details
Entrez Gene ID: | 77110 |
Gene Name: | GC-rich promoter binding protein 1-like 1 |
Gene Aliases: |
5330440M15Rik, BC002292 |
Location: |
Chr.4:116557157-116593882 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 9 - Exon 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Gpbp1l1 | NM_029868.2 | NP_084144.2 | ||
| AK037224.1 | BAC29763.1 | |||
| AK077148.1 | BAC36644.1 | |||
| AK133761.1 | BAE21827.1 | |||
| BC031428.1 | AAH31428.1 | |||
| BC066034.1 | AAH66034.1 |

Genomic Map