Assay Details
Target Gene Details
Entrez Gene ID: | 76299 |
Gene Name: | endoplasmic reticulum protein 44 |
Gene Aliases: |
1110001E24Rik, AI849526, AL033348, Txndc4 |
Location: |
Chr.4:48193331-48279601 on Build GRCm38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Erp44 | NM_029572.2 | 5 | 530 | NP_083848.1 |
| AK003217.1 | 5 | 499 | BAB22648.1 | |
| AK141377.1 | 5 | 488 | BAE24664.1 | |
| AK151497.1 | 5 | 531 | BAE30449.1 | |
| AK160113.1 | 5 | 513 | BAE35637.1 | |
| AK216793.1 | 2 | 141 | ||
| BC019558.1 | 5 | 437 | AAH19558.1 |

Genomic Map