Assay Details
Target Gene Details
Entrez Gene ID: | 76959 |
Gene Name: | charged multivesicular body protein 5 |
Gene Aliases: |
2210412K09Rik, AW545668 |
Location: |
Chr.4:40948553-40965302 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 3 - Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Chmp5 | NM_029814.1 | NP_084090.1 | ||
| AK008911.1 | BAB25962.1 | |||
| AK028446.1 | 1 | 2027 | ||
| AK147058.1 | BAE27642.1 | |||
| BC006947.1 | AAH06947.1 |
More Information
Set Membership: |
|

Genomic Map