Assay Details
Target Gene Details
Entrez Gene ID: | 76238 |
Gene Name: | glyoxylate reductase/hydroxypyruvate reductase |
Gene Aliases: |
1110059D05Rik, 6430629L09Rik |
Location: |
Chr.4:44981387-44992085 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Grhpr | NM_080289.1 | 2 | 182 | NP_525028.1 |
| AK004307.1 | 2 | 199 | ||
| AK018293.1 | 2 | 198 | ||
| AK168351.1 | 2 | 179 | BAE40288.1 | |
| AK172201.1 | 2 | 150 | BAE42878.1 | |
| AY113690.1 | 2 | 156 | AAM52985.1 | |
| BC010194.1 | 2 | 182 | AAH10194.1 |
More Information
Set Membership: |
|

Genomic Map