Assay Details
Target Gene Details
Entrez Gene ID: | 76238 |
Gene Name: | glyoxylate reductase/hydroxypyruvate reductase |
Gene Aliases: |
1110059D05Rik, 6430629L09Rik |
Location: |
Chr.4:44981387-44992085 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 7 - Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Grhpr | NM_080289.1 | NP_525028.1 | ||
| AK004307.1 | ||||
| AK018293.1 | ||||
| AK168351.1 | BAE40288.1 | |||
| AK172201.1 | BAE42878.1 | |||
| AY113690.1 | AAM52985.1 | |||
| BC010194.1 | AAH10194.1 |
More Information
Set Membership: |
|

Genomic Map