Assay Details
Target Gene Details
Entrez Gene ID: | 319636 |
Gene Name: | fibronectin type III and SPRY domain containing 1-like |
Gene Aliases: |
A230072O16Rik, Ccdc10, Csdufd1, Fsd1nl, OTTMUSG00000007002 |
Location: |
Chr.4:53629929-53707009 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Fsd1l | NM_001195284.1 | 1 | 68 | NP_001182213.1 |
| NM_176966.4 | 1 | 68 | NP_795940.1 | |
| AK038893.1 | 1 | 46 | BAC30161.1 | |
| AK140020.1 | 1 | 52 | BAE24213.1 | |
| BC130265.1 | ||||
| CJ115006.1 | 1 | 68 |
More Information
Set Membership: |
|

Genomic Map