Assay Details
Target Gene Details
Entrez Gene ID: | 230073 |
Gene Name: | DEAD (Asp-Glu-Ala-Asp) box polypeptide 58 |
Gene Aliases: |
6430573D20Rik, C330021E21, RIG-I, RLR-1 |
Location: |
Chr.4:40203777-40239825 on Build GRCm38 |
Assay Gene Location: | Within Exon 12 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ddx58 | NM_172689.3 | 12 | 1837 | NP_766277.3 |
| AK049305.1 | 3 | 428 | BAC33670.1 | |
| AK052595.1 | 8 | 1091 | ||
| AK128929.1 | 12 | 1772 | ||
| AK155930.1 | 12 | 1773 | BAE33508.1 | |
| AY553221.1 | 12 | 1713 | AAS59532.1 | |
| BC027369.1 | 12 | 1816 | ||
| BC106095.1 | 12 | 1757 | AAI06096.1 | |
| BC127163.1 | 12 | 1703 | ||
| BC152540.1 | 12 | 1705 |

Genomic Map