Assay Details
Target Gene Details
Entrez Gene ID: | 230073 |
Gene Name: | DEAD (Asp-Glu-Ala-Asp) box polypeptide 58 |
Gene Aliases: |
6430573D20Rik, C330021E21, RIG-I, RLR-1 |
Location: |
Chr.4:40203777-40239825 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 4 - Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ddx58 | NM_172689.3 | NP_766277.3 | ||
| AK052871.1 | BAC35183.1 | |||
| AK078287.1 | BAC37205.1 | |||
| AK087261.1 | 4 | 706 | BAC39830.1 | |
| AK128929.1 | ||||
| AK155930.1 | BAE33508.1 | |||
| AY553221.1 | AAS59532.1 | |||
| BC027369.1 | ||||
| BC106095.1 | AAI06096.1 | |||
| BC127163.1 | ||||
| BC152540.1 |

Genomic Map