Assay Details
Target Gene Details
Entrez Gene ID: | 74246 |
Gene Name: | galactose-4-epimerase, UDP |
Gene Aliases: |
2310002A12Rik, AI323962 |
Location: |
Chr.4:135963681-135968178 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 5 - Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Gale | NM_178389.3 | NP_848476.1 | ||
| AK009083.1 | ||||
| AK146184.1 | ||||
| BC027438.1 | AAH27438.1 |
More Information
Set Membership: |
|

Genomic Map