Assay Details
Target Gene Details
Entrez Gene ID: | 15206 |
Gene Name: | hairy and enhancer of split 2 (Drosophila) |
Gene Aliases: |
HES-2 |
Location: |
Chr.4:152158867-152162469 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Hes2 | NM_008236.4 | NP_032262.2 | ||
| AK053792.1 | BAC35525.1 | |||
| BC138112.1 | 2 | 434 |
More Information
Set Membership: |
|

Genomic Map