Assay Details
Target Gene Details
Entrez Gene ID: | 15208 |
Gene Name: | hairy and enhancer of split 5 (Drosophila) |
Gene Aliases: |
bHLHb38 |
Location: |
Chr.4:154958680-154962372 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Hes5 | NM_010419.4 | 1 | 57 | NP_034549.1 |
| AK078170.1 | 1 | 60 | BAC37159.1 | |
| AK087508.1 | 1 | 58 | BAC39904.1 | |
| AK139390.1 | 1 | 59 | BAE23990.1 | |
| AK156545.1 | 1 | 59 | BAE33752.1 | |
| BC099863.1 | AAH99863.1 | |||
| BC099864.1 | AAH99864.1 | |||
| BC099865.1 | AAH99865.1 | |||
| BC103539.1 | AAI03540.1 |
More Information
Set Membership: |
|

Genomic Map