Assay Details
Target Gene Details
Entrez Gene ID: | 68917 |
Gene Name: | histidine triad nucleotide binding protein 2 |
Gene Aliases: |
1190005L05Rik |
Location: |
Chr.4:43654227-43656486 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Hint2 | NM_026871.1 | NP_081147.1 | ||
| AF356874.1 | AAM00220.1 | |||
| AK004497.1 | BAB23334.1 | |||
| AK170135.1 | ||||
| BC086940.1 | AAH86940.1 |
More Information
Set Membership: |
|

Genomic Map