Assay Details
Target Gene Details
Entrez Gene ID: | 212377 |
Gene Name: | MMS22-like, DNA repair protein |
Gene Aliases: |
F730047E07Rik, Gm134 |
Location: |
Chr.4:24496444-24602950 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 9 - Exon 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Mms22l | NM_199467.2 | NP_955761.2 | ||
| AK089531.1 | ||||
| AK144799.1 | ||||
| AK166072.1 | ||||
| AY512923.1 | AAR87794.1 | |||
| BC051674.1 | AAH51674.1 | |||
| BC137910.1 | ||||
| BC145028.1 |

Genomic Map