Assay Details
Target Gene Details
Entrez Gene ID: | 230125 |
Gene Name: | solute carrier family 25, member 51 |
Gene Aliases: |
9130208E07Rik, D130005A03Rik, Gm138, Mcart1 |
Location: |
Chr.4:45395923-45408783 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Slc25a51 | NM_001009949.3 | NP_001009949.3 | ||
| AK020275.1 | ||||
| AK146788.1 | BAE27432.1 | |||
| AK150362.1 | BAE29497.1 | |||
| AK151688.1 | BAE30612.1 | |||
| AK152171.1 | BAE31003.1 | |||
| AK156168.1 | BAE33610.1 | |||
| AK156342.1 | ||||
| AK164726.1 | ||||
| AK165253.1 | BAE38106.1 | |||
| AK172225.1 | 1 | 113 | BAE42892.1 |

Genomic Map