Assay Details
Target Gene Details
Entrez Gene ID: | 17480 |
Gene Name: | myeloproliferative leukemia virus oncogene |
Gene Aliases: |
CD110, TPO-R, c-mpl, hlb219 |
Location: |
Chr.4:118442412-118457513 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 7 - Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Mpl | NM_001122949.2 | NP_001116421.1 | ||
| NM_001285496.1 | NP_001272425.1 | |||
| NM_001285497.1 | NP_001272426.1 | |||
| NM_010823.3 | NP_034953.2 | |||
| AF360122.1 | AAK52492.1 | |||
| AK041166.1 | BAC30846.1 | |||
| BC103513.1 | AAI03514.1 | |||
| BC103515.1 | AAI03516.1 | |||
| BC146612.1 | ||||
| BC146613.1 | ||||
| X73677.1 | CAA52031.1 | |||
| Z22649.1 | CAA80365.1 |
More Information
Set Membership: |
|

Genomic Map