Assay Details
Target Gene Details
Entrez Gene ID: | 72656 |
Gene Name: | integrator complex subunit 8 |
Gene Aliases: |
2810013E07Rik, AV063769, D130008D20Rik |
Location: |
Chr.4:11199158-11254266 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 23 - Intron 24 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ints8 | NM_001159595.1 | NP_001153067.1 | ||
| NM_178112.5 | NP_835213.3 | |||
| AK039325.1 | ||||
| AK049513.1 | BAC33786.1 | |||
| AK140829.1 | BAE24491.1 | |||
| AK164951.1 | BAE37977.1 | |||
| AK169552.1 | BAE41225.1 | |||
| AK204383.1 | ||||
| BC049092.1 | AAH49092.1 | |||
| BC050192.1 | AAH50192.1 | |||
| BC057618.1 | AAH57618.1 |
More Information
Set Membership: |
|

Genomic Map