Assay Details
Target Gene Details
Entrez Gene ID: | 69902 |
Gene Name: | MRT4, mRNA turnover 4, homolog (S. cerevisiae) |
Gene Aliases: |
2610012O22Rik, Mg684, Mrt4 |
Location: |
Chr.4:139347440-139352576 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 5 - Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Mrto4 | NM_001290810.1 | NP_001277739.1 | ||
| NM_023536.3 | NP_076025.1 | |||
| AK011387.1 | BAB27585.1 | |||
| AK171445.1 | 2 | 2055 | ||
| AK192789.1 | ||||
| BC005734.1 | AAH05734.1 | |||
| BC138221.1 | ||||
| BC145205.1 |

Genomic Map