Assay Details
Target Gene Details
Entrez Gene ID: | 100039672 |
Gene Name: | microseminoprotein, prostate associated |
Gene Aliases: |
OTTMUSG00000007017, Psmp |
Location: |
Chr.4:43583216-43584494 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Msmp | NM_001099314.1 | 2 | 414 | NP_001092784.1 |
Target Gene Details
Entrez Gene ID: | 242406 |
Gene Name: | RGP1 retrograde golgi transport homolog (S. cerevisiae) |
Gene Aliases: |
1110029E03Rik, AI527196, F730001J03, mKIAA0258 |
Location: |
Chr.4:43578735-43587487 on Build GRCm38 |
Assay Gene Location: | Within Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Rgp1 | NM_172866.3 | 9 | 2245 | NP_766454.1 |
| AK129103.1 | 9 | 2031 | BAC97913.1 | |
| AK170117.1 | BAE41575.1 | |||
| BC086614.1 | 9 | 2051 | AAH86614.1 |
More Information
Set Membership: |
|

Genomic Map