Assay Details
Target Gene Details
Entrez Gene ID: | 100126227 |
Gene Name: | predicted gene 13031 |
Gene Aliases: |
OTTMUSG00000010081 |
Location: |
Chr.4:140947666-140957902 on Build GRCm38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Gm13031 | NR_045911.1 | |||
| AK040418.1 | ||||
| BE651383.1 |
Target Gene Details
Entrez Gene ID: | 18600 |
Gene Name: | peptidyl arginine deiminase, type II |
Gene Aliases: |
Pdi, Pdi2, mKIAA0994 |
Location: |
Chr.4:140906334-140952586 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 14 - Exon 15 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Padi2 | NM_008812.2 | NP_032838.2 | ||
| AK033760.1 | ||||
| AK148156.1 | BAE28381.1 | |||
| AK154814.1 | ||||
| AK158382.1 | ||||
| AK168685.1 | ||||
| AK171275.1 | BAE42360.1 | |||
| BC040350.1 | AAH40350.1 | |||
| BC049947.1 | AAH49947.1 | |||
| BC131636.1 | ||||
| D16580.1 | BAA04012.1 |
More Information
Set Membership: |
|

Genomic Map