Assay Details
Target Gene Details
Entrez Gene ID: | 230828 |
Gene Name: | interleukin 22 receptor, alpha 1 |
Gene Aliases: |
9130219A07Rik, ENSMUSG00000073746, IL-22R, Il22r |
Location: |
Chr.4:135728159-135755383 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Il22ra1 | NM_178257.2 | 3 | 318 | NP_839988.1 |
| AK141289.1 | 3 | 319 | BAE24639.1 | |
| AY103454.1 | 3 | 257 | AAM52222.1 | |
| BC140972.1 | 3 | 324 | ||
| BC140973.1 | 3 | 324 |
More Information
Set Membership: |
|

Genomic Map