Assay Details
Target Gene Details
Entrez Gene ID: | 319865 |
Gene Name: | RIKEN cDNA E130114P18 gene |
Gene Aliases: |
- |
Location: |
Chr.4:97567875-97584591 on Build GRCm38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| E130114P18Rik | NR_015513.2 | |||
| AK082601.1 | ||||
| AK086602.1 |
Target Gene Details
Entrez Gene ID: | 18027 |
Gene Name: | nuclear factor I/A |
Gene Aliases: |
1110047K16Rik, 9430022M17Rik, CTF, NF1-A, NF1A |
Location: |
Chr.4:97581717-98118876 on Build GRCm38 |
Assay Gene Location: | Overlaps - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Nfia | NM_001122952.1 | NP_001116424.1 | ||
| AK139357.1 | BAE23974.1 | |||
| D90173.1 | 1 | 63 | BAA14203.1 |
More Information
Set Membership: |
|

Genomic Map