Assay Details
Target Gene Details
Entrez Gene ID: | 80509 |
Gene Name: | mediator complex subunit 8 |
Gene Aliases: |
2210021A15Rik, AB041805, ARC32 |
Location: |
Chr.4:118409298-118415824 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 4 - Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Med8 | NM_001290688.1 | NP_001277617.1 | ||
| NM_020000.3 | NP_064384.2 | |||
| NM_173719.3 | NP_776067.1 | |||
| AB041805.1 | BAA95113.1 | |||
| AK005422.1 | BAB24016.1 | |||
| AK008759.1 | BAB25879.1 | |||
| AK011080.1 | BAB27384.1 | |||
| AK147032.1 | BAE27624.1 | |||
| AK148025.1 | BAE28295.1 | |||
| BC002315.1 | AAH02315.1 | |||
| BC021870.1 | AAH21870.1 |
More Information
Set Membership: |
|

Genomic Map