Assay Details
Target Gene Details
Entrez Gene ID: | 74761 |
Gene Name: | matrix-remodelling associated 8 |
Gene Aliases: |
1200013A08Rik, 1700095D18Rik, AI131686, Asp3, Dicam |
Location: |
Chr.4:155839680-155844102 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 7 - Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Mxra8 | NM_024263.4 | NP_077225.4 | ||
| AB040488.1 | BAB68501.1 | |||
| AB052097.1 | BAD05133.1 | |||
| AK004732.1 | BAB23514.1 | |||
| AK195482.1 | ||||
| BC026438.1 | AAH26438.1 | |||
| EU849596.1 | ||||
| EU849597.1 |

Genomic Map