Assay Details
Target Gene Details
Entrez Gene ID: | 12695 |
Gene Name: | InaD-like (Drosophila) |
Gene Aliases: |
Cipp, Patj |
Location: |
Chr.4:98395534-98719603 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 14 - Intron 14 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Inadl | NM_001005784.1 | NP_001005784.1 | ||
| NM_172696.2 | NP_766284.2 | |||
| AK050545.1 | ||||
| AK147866.1 | BAE28189.1 | |||
| AK153597.1 | BAE32113.1 | |||
| AK156166.1 | BAE33609.1 | |||
| BC050846.1 | AAH50846.1 | |||
| BC062194.1 | AAH62194.1 | |||
| BC082787.1 | AAH82787.1 | |||
| BC141404.1 |

Genomic Map