Assay Details
Target Gene Details
Entrez Gene ID: | 12695 |
Gene Name: | InaD-like (Drosophila) |
Gene Aliases: |
Cipp, Patj |
Location: |
Chr.4:98395534-98719603 on Build GRCm38 |
Assay Gene Location: | Within Exon 22 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Inadl | NM_172696.2 | 22 | 3190 | NP_766284.2 |
| AK050545.1 | 18 | 2166 | ||
| AK077268.1 | 9 | 1472 | BAC36720.1 | |
| AK147866.1 | 22 | 3139 | BAE28189.1 | |
| BC082787.1 | 11 | 1583 | AAH82787.1 | |
| BC141404.1 | 22 | 3166 |

Genomic Map