Assay Details
Target Gene Details
Entrez Gene ID: | 212390 |
Gene Name: | kelch-like 32 |
Gene Aliases: |
6430524H05Rik, D4Ertd389e, Gm1356, mKIAA1900 |
Location: |
Chr.4:24614352-24851110 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 7 - Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Klhl32 | NM_001033531.3 | NP_001028703.2 | ||
| NM_001163020.1 | NP_001156492.1 | |||
| AK134944.1 | BAE22350.1 | |||
| BC141239.1 |

Genomic Map