Assay Details
Target Gene Details
Entrez Gene ID: | 26970 |
Gene Name: | phospholipase A2, group IIE |
Gene Aliases: |
- |
Location: |
Chr.4:138877911-138882817 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 1 - Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Pla2g2e | NM_012044.2 | NP_036174.1 | ||
| AF112984.1 | AAF22290.1 | |||
| AF166098.1 | AAF04499.1 |
More Information
Set Membership: |
|

Genomic Map