Assay Details
Target Gene Details
Entrez Gene ID: | 67103 |
Gene Name: | prostaglandin reductase 1 |
Gene Aliases: |
2510002C21Rik, Ltb4dh, Zadh3 |
Location: |
Chr.4:58965590-58987131 on Build GRCm38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ptgr1 | NM_025968.3 | 5 | 350 | NP_080244.1 |
| AK010888.1 | 5 | 385 | BAB27248.1 | |
| AK011962.1 | 5 | 372 | BAB27941.1 | |
| AK035425.1 | 5 | 351 | BAC29060.1 | |
| AK134440.1 | 5 | 383 | BAE22145.1 | |
| AK166835.1 | 5 | 350 | BAE39057.1 | |
| BC014865.1 | 5 | 332 | AAH14865.1 | |
| CT010267.1 | 4 | 299 | CAJ18475.1 |
More Information
Set Membership: |
|

Genomic Map