Assay Details
Target Gene Details
Entrez Gene ID: | 12695 |
Gene Name: | InaD-like (Drosophila) |
Gene Aliases: |
Cipp, Patj |
Location: |
Chr.4:98395534-98719603 on Build GRCm38 |
Assay Gene Location: | Within Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Inadl | NM_001005784.1 | 6 | 717 | NP_001005784.1 |
| NM_172696.2 | 6 | 717 | NP_766284.2 | |
| AK050545.1 | 6 | 689 | ||
| AK147866.1 | 6 | 666 | BAE28189.1 | |
| BC050846.1 | 6 | 651 | AAH50846.1 | |
| BC141404.1 | 6 | 693 |

Genomic Map