Assay Details
Target Gene Details
Entrez Gene ID: | 545622 |
Gene Name: | protein tyrosine phosphatase, non-receptor type 3 |
Gene Aliases: |
9530011I20Rik, PTP-H1, PTPCL |
Location: |
Chr.4:57190841-57339046 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 24 - Exon 24 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ptpn3 | NM_011207.2 | NP_035337.2 |
More Information
Set Membership: |
|

Genomic Map