Assay Details
Target Gene Details
Entrez Gene ID: | 230316 |
Gene Name: | multiple EGF-like-domains 9 |
Gene Aliases: |
4933405H16Rik, 9630025C07, Egfl5, mKIAA0818 |
Location: |
Chr.4:70431927-70534928 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 3 - Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Megf9 | NM_172694.2 | NP_766282.1 | ||
| AK035987.1 | BAC29269.1 | |||
| AK039319.1 | BAC30317.1 | |||
| BC088994.1 | ||||
| BC138128.1 | ||||
| BC138129.1 |

Genomic Map