Assay Details
Target Gene Details
Entrez Gene ID: | 19266 |
Gene Name: | protein tyrosine phosphatase, receptor type, D |
Gene Aliases: |
1110002J03Rik, 3000002J10Rik, B230219D21Rik, R-PTP-delta |
Location: |
Chr.4:75941237-78211895 on Build GRCm38 |
Assay Gene Location: | Within Exon 31 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ptprd | NM_011211.3 | 31 | 3970 | NP_035341.2 |
| AF326559.1 | 13 | 2523 | AAL37405.1 | |
| AF326560.1 | 13 | 2523 | AAL37406.1 | |
| AK051809.1 | 8 | 1031 | BAE43348.1 | |
| BC025145.1 | 7 | 815 | AAH25145.1 | |
| BC141409.1 | 21 | 3149 | ||
| D13903.1 | 13 | 2926 | BAA03003.1 | |
| Z23051.1 | CAA80586.1 |
More Information
Set Membership: |
|

Genomic Map