Assay Details
Target Gene Details
Entrez Gene ID: | 21942 |
Gene Name: | tumor necrosis factor receptor superfamily, member 9 |
Gene Aliases: |
4-1BB, A930040I11Rik, AA408498, AI325004, CDw137, Cd137, ILA, Ly63 |
Location: |
Chr.4:150920155-150946104 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 2 - Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Tnfrsf9 | NM_001077508.1 | NP_001070976.1 | ||
| NM_001077509.1 | NP_001070977.1 | |||
| NM_011612.2 | NP_035742.1 | |||
| AK019885.1 | ||||
| AK154625.1 | BAE32724.1 | |||
| AK170670.1 | 1 | 724 | ||
| BC028507.1 | AAH28507.1 | |||
| BY196142.1 | ||||
| BY206772.1 |
More Information
Set Membership: |
|

Genomic Map