Assay Details
Target Gene Details
Entrez Gene ID: | 69934 |
Gene Name: | tRNA methyltransferase 10B |
Gene Aliases: |
2610042J10Rik, Rg9mtd3, Rnmtd3 |
Location: |
Chr.4:45297071-45316131 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 6 - Intron 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Trmt10b | NM_027266.4 | NP_081542.2 | ||
| AK011746.1 | BAB27815.1 | |||
| AK030465.1 | BAC26974.1 | |||
| BC019428.1 | AAH19428.1 |
More Information
Set Membership: |
|

Genomic Map