Assay Details
Target Gene Details
Entrez Gene ID: | 14211 |
Gene Name: | structural maintenance of chromosomes 2 |
Gene Aliases: |
5730502P04Rik, AI255214, AW545314, CAP-E, CAPE, Fin16, SMC-2, Smc2l1 |
Location: |
Chr.4:52439221-52488365 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 13 - Intron 13 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Smc2 | NM_001301412.1 | NP_001288341.1 | ||
| NM_008017.4 | NP_032043.3 | |||
| AJ534939.1 | CAD59182.1 | |||
| AK028233.1 | ||||
| AK145341.1 | BAE26376.1 | |||
| AK152090.1 | ||||
| BC094380.1 | AAH94380.1 |

Genomic Map