Assay Details
Target Gene Details
Entrez Gene ID: | 230157 |
Gene Name: | transmembrane protein with EGF-like and two follistatin-like domains 1 |
Gene Aliases: |
A830033E11, M7365, Tr1 |
Location: |
Chr.4:48585114-48663131 on Build GRCm38 |
Assay Gene Location: | Within Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Tmeff1 | NM_021436.2 | 6 | 878 | NP_067411.1 |
| AJ400622.1 | 6 | 547 | CAB90827.1 | |
| AK043792.1 | 4 | 254 | BAC31655.1 | |
| AK079633.1 | 4 | 234 | BAC37709.1 | |
| BC057598.1 | 6 | 878 | AAH57598.1 |
More Information
Set Membership: |
|

Genomic Map