Assay Details
Target Gene Details
Entrez Gene ID: | 242406 |
Gene Name: | RGP1 retrograde golgi transport homolog (S. cerevisiae) |
Gene Aliases: |
1110029E03Rik, AI527196, F730001J03, mKIAA0258 |
Location: |
Chr.4:43578735-43587487 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 8 - Intron 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Rgp1 | NM_172866.3 | NP_766454.1 | ||
| AK089288.1 | BAC40830.1 | |||
| AK129103.1 | BAC97913.1 | |||
| AK170117.1 | BAE41575.1 | |||
| BC086614.1 | AAH86614.1 |

Genomic Map