Assay Details
Target Gene Details
Entrez Gene ID: | 622404 |
Gene Name: | coiled-coil domain containing 107 |
Gene Aliases: |
- |
Location: |
Chr.4:43493340-43495921 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 3 - Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ccdc107 | NM_001037913.2 | NP_001033002.1 | ||
| AK002915.1 | BAB22454.1 | |||
| AK004044.1 | BAB23140.1 | |||
| BC027414.1 | AAH27414.1 |

Genomic Map