Assay Details
Target Gene Details
Entrez Gene ID: | 76421 |
Gene Name: | RIKEN cDNA 1700028K03 gene |
Gene Aliases: |
- |
Location: |
Chr.5:107534711-107551542 on Build GRCm38 |
Assay Gene Location: | Overlaps - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| 1700028K03Rik | NM_175241.1 | NP_780450.1 | ||
| NM_182745.2 | NP_877422.1 | |||
| AK006463.1 | BAC25144.1 | |||
| AK032886.1 |

Genomic Map