Assay Details
Target Gene Details
Entrez Gene ID: | 654318 |
Gene Name: | RIKEN cDNA C530005A16 gene |
Gene Aliases: |
D930044c15rik |
Location: |
Chr.4:116589733-116597630 on Build GRCm38 |
Assay Gene Location: | Overlaps - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| C530005A16Rik | NR_029450.1 | |||
| AK035052.1 | ||||
| AK039789.1 | ||||
| AK086654.1 | BAC39711.1 | |||
| AK140096.1 |
Target Gene Details
Entrez Gene ID: | 622665 |
Gene Name: | coiled-coil domain containing 17 |
Gene Aliases: |
- |
Location: |
Chr.4:116596730-116600266 on Build GRCm38 |
Assay Gene Location: | Within Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ccdc17 | NM_001037916.3 | NP_001033005.1 | ||
| AK029257.1 | BAC26359.1 | |||
| AK076542.1 | ||||
| BC150874.1 | ||||
| BC172023.1 |
More Information
Set Membership: |
|

Genomic Map